Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFS4
Entrez Gene ID
327680
Gene Name
NADH:ubiquinone oxidoreductase subunit S4
Gene Aliases
-
Chromosome Location
Chr.20: 25400369 - 25515962 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175800.2 NP_786994.1 4 - 5 443 71
GenBank mRNA X63215.1 - - 443 71

View Full Product Details