Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAG20
Entrez Gene ID
337914
Gene Name
pregnancy-associated glycoprotein 20
Gene Aliases
-
Chromosome Location
Chr.29: 38428593 - 38437719 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176629.1 NP_788802.1 2 - 3 196 83
GenBank mRNA AF192337.1 - 2 - 3 196 83

View Full Product Details