Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIB2
Entrez Gene ID
352960
Gene Name
tribbles pseudokinase 2
Gene Aliases
TRB-2, TRB2
Chromosome Location
Chr.11: 85237101 - 85266651 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178317.3 NP_847887.2 1 - 2 269 73
GenBank mRNA AY247741.1 - 1 - 2 764 73
AY360147.1 - 1 - 2 269 73
BC123595.1 - 1 - 2 811 73

View Full Product Details