Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCA3
Entrez Gene ID
281694
Gene Name
chloride channel regulator 3
Gene Aliases
-
Chromosome Location
Chr.3: 57420369 - 57445306 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181018.2 NP_851361.1 14 - 15 2441 136
GenBank mRNA AF001261.1 - 14 - 15 2441 136

View Full Product Details