Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TKDP4
Entrez Gene ID
404075
Gene Name
trophoblast Kunitz domain protein 4
Gene Aliases
-
Chromosome Location
Chr.13: 74106679 - 74114983 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205775.1 NP_991344.1 4 - 5 340 69
GenBank mRNA AF241779.1 - 4 - 5 340 69

View Full Product Details