Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKDP1
Entrez Gene ID
404076
Gene Name
trophoblast Kunitz domain protein 1
Gene Aliases
-
Chromosome Location
Chr.13: 74075296 - 74092083 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205776.1 NP_991345.1 5 - 6 435 101
GenBank mRNA AF241776.1 - 5 - 6 435 101

View Full Product Details