Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP16
Entrez Gene ID
506421
Gene Name
NOP16 nucleolar protein
Gene Aliases
HSPC111
Chromosome Location
Chr.7: 37851239 - 37855996 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014856.1 NP_001014856.1 2 - 3 252 91
GenBank mRNA BC134466.1 - 2 - 3 275 91
BT021024.1 - 2 - 3 252 91

View Full Product Details