Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF34
Entrez Gene ID
506764
Gene Name
ring finger protein 34
Gene Aliases
-
Chromosome Location
Chr.17: 53771759 - 53798675 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014858.1 NP_001014858.1 3 - 4 715 140
GenBank mRNA BC110246.1 - 4 - 5 969 140
BT020924.1 - 3 - 4 715 140

View Full Product Details