Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
513211
Gene Name
HSPB1 associated protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 67064196 - 67121243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014911.1 NP_001014911.1 6 - 7 916 75
GenBank mRNA BT021854.1 - 6 - 7 916 75

View Full Product Details