Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM45
Entrez Gene ID
539091
Gene Name
tripartite motif containing 45
Gene Aliases
-
Chromosome Location
Chr.3: 26099795 - 26109125 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014952.1 NP_001014952.1 2 - 3 1817 63
GenBank mRNA BT021203.1 - 2 - 3 1817 63

View Full Product Details