Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN2L
Entrez Gene ID
539289
Gene Name
lectin, mannose binding 2 like
Gene Aliases
-
Chromosome Location
Chr.11: 2683655 - 2704108 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014953.1 NP_001014953.1 8 - 9 948 106
GenBank mRNA BC105565.1 - 7 - 8 106
BT021149.1 - 8 - 9 948 106

View Full Product Details