Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THSD1
Entrez Gene ID
541228
Gene Name
thrombospondin type 1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 21651368 - 21679255 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014967.1 NP_001014967.1 3 - 4 1188 72
GenBank mRNA BC140588.1 - 3 - 4 1198 72
BT021161.1 - 3 - 4 1188 72

View Full Product Details