Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL40
Entrez Gene ID
514526
Gene Name
kelch like family member 40
Gene Aliases
KBTBD5
Chromosome Location
Chr.22: 14817188 - 14824003 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015598.1 NP_001015598.1 5 - 6 1798 115
GenBank mRNA BT021600.1 - 5 - 6 1798 115

View Full Product Details