Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDIN1
Entrez Gene ID
538758
Gene Name
CDAN1 interacting nuclease 1
Gene Aliases
C10H15orf41, C15orf41, HH114
Chromosome Location
Chr.10: 32243714 - 32477173 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015668.1 NP_001015668.1 5 - 6 497 76
GenBank mRNA BC134779.1 - 2 - 3 103 76
BC142022.1 - 5 - 6 577 76
BT020842.1 - 5 - 6 497 76

View Full Product Details