Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF23
Entrez Gene ID
539774
Gene Name
PHD finger protein 23
Gene Aliases
-
Chromosome Location
Chr.19: 26948834 - 26953195 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024570.1 NP_001019741.1 4 - 5 1116 71
GenBank mRNA BC142215.1 - 4 - 5 1155 71
BT021849.1 - 4 - 5 1116 71
BT030481.1 - 4 - 5 1149 71

View Full Product Details