The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034261.2 | NP_001029433.1 | 1 - 2 | 218 | 108 |
GenBank mRNA | BC103175.1 | - | 1 - 2 | 219 | 108 |