Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICOS
Entrez Gene ID
507026
Gene Name
inducible T cell costimulator
Gene Aliases
-
Chromosome Location
Chr.2: 92144638 - 92169894 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034275.1 NP_001029447.1 2 - 3 407 136
GenBank mRNA BT021638.1 - 2 - 3 407 136

View Full Product Details