Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD7
Entrez Gene ID
512456
Gene Name
proteasome 26S subunit, non-ATPase 7
Gene Aliases
-
Chromosome Location
Chr.18: 36981892 - 36989873 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034423.2 NP_001029595.1 3 - 4 364 143
GenBank mRNA BC103437.1 - 3 - 4 365 143
BT029898.1 - 3 - 4 331 143

View Full Product Details