Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAIP2
Entrez Gene ID
536619
Gene Name
poly(A) binding protein interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.7: 50627934 - 50645912 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034636.2 NP_001029808.1 1 - 2 149 73
GenBank mRNA BC102885.1 - 1 - 2 151 73

View Full Product Details