Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB14
Entrez Gene ID
509431
Gene Name
ankyrin repeat and SOCS box containing 14
Gene Aliases
DNAH12
Chromosome Location
Chr.22: 43831575 - 43855360 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035070.2 NP_001030242.2 6 - 7 755 96
GenBank mRNA BC148874.1 - 6 - 7 756 96
BT021867.1 - 1 - 2 184 96

View Full Product Details