Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRUNE1
Entrez Gene ID
524973
Gene Name
prune exopolyphosphatase 1
Gene Aliases
PRUNE
Chromosome Location
Chr.3: 19746347 - 19766243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035352.1 NP_001030429.1 5 - 6 675 86
GenBank mRNA BC142289.1 - 5 - 6 690 86
BT020783.1 - 5 - 6 675 86

View Full Product Details