Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYL1
Entrez Gene ID
281725
Gene Name
crystallin lambda 1
Gene Aliases
-
Chromosome Location
Chr.12: 35929011 - 35985473 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174293.2 NP_776718.1 5 - 6 692 73
GenBank mRNA AF480862.1 - 5 - 6 692 73
BC140654.1 - 5 - 6 694 73

View Full Product Details