Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC1A5
Entrez Gene ID
282355
Gene Name
solute carrier family 1 member 5
Gene Aliases
ASCT2
Chromosome Location
Chr.18: 53864326 - 53875875 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174601.2 NP_777026.1 4 - 5 1437 60
GenBank mRNA AY039236.1 - 4 - 5 1437 60
BC123803.1 - 4 - 5 1026 60
LC005614.1 - 4 - 5 822 60

View Full Product Details