Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STATH
Entrez Gene ID
286765
Gene Name
statherin
Gene Aliases
-
Chromosome Location
Chr.6: 85497254 - 85506604 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174752.3 NP_777177.1 2 - 3 77 86
GenBank mRNA AY154893.1 - 2 - 3 77 86

View Full Product Details