Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADGRE5
Entrez Gene ID
338066
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97
Chromosome Location
Chr.7: 11349799 - 11371529 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176661.1 NP_788834.1 2 - 3 93 70
GenBank mRNA AJ416058.1 - 2 - 3 93 70
AJ627191.1 - 2 - 3 145 70
BC149039.1 - 2 - 3 144 70

View Full Product Details