Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FDPS
Entrez Gene ID
281156
Gene Name
farnesyl diphosphate synthase
Gene Aliases
-
Chromosome Location
Chr.3: 15311071 - 15320374 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177497.2 NP_803463.1 2 - 3 294 75
GenBank mRNA AF461050.1 - 2 - 3 294 75
BC111256.1 - 3 - 4 1114 75
BC120218.1 - 3 - 4 356 75
BC149572.1 - 3 - 4 415 75

View Full Product Details