Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GHRH
Entrez Gene ID
281191
Gene Name
growth hormone releasing hormone
Gene Aliases
-
Chromosome Location
Chr.13: 66207015 - 66216486 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178325.1 NP_847895.1 2 - 3 164 124

View Full Product Details