Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ADAMTS4
Entrez Gene ID
286806
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
-
Chromosome Location
Chr.3: 8303802 - 8311858 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181667.1 NP_858053.1 8 - 9 2488 59
GenBank mRNA AF516915.1 - 8 - 9 2488 59
BC148059.1 - 8 - 9 2500 59

View Full Product Details