Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COX4I1
Entrez Gene ID
281090
Gene Name
cytochrome c oxidase subunit 4I1
Gene Aliases
COX4
Chromosome Location
Chr.18: 11758912 - 11767079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001439.3 NP_001001439.1 2 - 3 431 67
GenBank mRNA BC102733.1 - 2 - 3 432 67
BT021029.1 - 2 - 3 180 67

Gene Symbol
COXP1
Entrez Gene ID
281710
Gene Name
cytochrome c oxidase, pseudogene
Gene Aliases
-
Chromosome Location
Chr.5: 110539192 - 110539865 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA Y14076.1 - - 284 67

View Full Product Details