Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL2
Entrez Gene ID
444879
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Gene Aliases
SIAT4B
Chromosome Location
Chr.18: 1678017 - 1725467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002892.2 NP_001002892.1 3 - 4 1204 93
GenBank mRNA AJ748841.1 - 3 - 4 716 93
BC105252.1 - 3 - 4 1204 93

View Full Product Details