Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RTCB
Entrez Gene ID
525106
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
C22orf28, C5H22orf28, HSPC117
Chromosome Location
Chr.5: 71017591 - 71038959 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015631.2 NP_001015631.1 2 - 3 209 118
GenBank mRNA BC104498.1 - 2 - 3 209 118
BT020831.1 - 2 - 3 222 118

View Full Product Details