Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL1A1
Entrez Gene ID
282187
Gene Name
collagen type I alpha 1 chain
Gene Aliases
-
Chromosome Location
Chr.19: 36457701 - 36475647 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034039.2 NP_001029211.1 27 - 28 1993 77
GenBank mRNA BC105184.1 - 27 - 28 1994 77

View Full Product Details