Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB5IF
Entrez Gene ID
512377
Gene Name
RAB5 interacting factor
Gene Aliases
C13H20orf24, RIP5
Chromosome Location
Chr.13: 65704921 - 65712336 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034421.2 NP_001029593.1 1 - 2 296 88
GenBank mRNA BC102069.1 - 1 - 2 297 88

View Full Product Details