Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGN3
Entrez Gene ID
515652
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
-
Chromosome Location
Chr.9: 18779302 - 18814816 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034504.2 NP_001029676.1 5 - 6 337 73
GenBank mRNA BC103089.1 - 5 - 6 338 73

View Full Product Details