Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMTOR2
Entrez Gene ID
616375
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Gene Aliases
MAPBPIP, ROBLD3
Chromosome Location
Chr.3: 14709191 - 14712337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034791.1 NP_001029963.1 2 - 3 282 98
GenBank mRNA BC102146.1 - 2 - 3 282 98

View Full Product Details