Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C7H5orf15
Entrez Gene ID
514781
Gene Name
chromosome 7 C5orf15 homolog
Gene Aliases
-
Chromosome Location
Chr.7: 45648089 - 45661066 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035299.1 NP_001030376.1 2 - 3 683 138
GenBank mRNA BC102841.1 - 2 - 3 683 138

View Full Product Details