Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ENKD1
Entrez Gene ID
524652
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, C18H16orf48
Chromosome Location
Chr.18: 35216694 - 35220196 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035350.2 NP_001030427.1 6 - 7 858 140
GenBank mRNA BC102534.1 - 6 - 7 860 140

View Full Product Details