The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037485.2 | NP_001032562.1 | 1 - 2 | 98 | 143 |
GenBank mRNA | BC108247.1 | - | 1 - 2 | 100 | 143 |