Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOF
Entrez Gene ID
617894
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
-
Chromosome Location
Chr.17: 53499932 - 53523522 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040602.2 NP_001035692.1 3 - 4 367 66
GenBank mRNA BC103016.1 - 3 - 4 368 66

View Full Product Details