Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN5
Entrez Gene ID
614076
Gene Name
MORN repeat containing 5
Gene Aliases
C11H9orf18, C9orf18
Chromosome Location
Chr.11: 93046908 - 93081440 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076310.2 NP_001069778.1 1 - 2 105 121
GenBank mRNA BC109519.1 - 1 - 2 106 121

View Full Product Details