Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS29
Entrez Gene ID
286884
Gene Name
ribosomal protein S29
Gene Aliases
-
Chromosome Location
Chr.10: 42627498 - 42629350 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174804.3 NP_777229.1 1 - 1 96 85
GenBank mRNA BC102702.1 - 1 - 1 96 85
U66372.1 - 1 - 2 73 85

View Full Product Details