Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF2BP
Entrez Gene ID
617142
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
Chromosome Location
Chr.1: 143641658 - 143729079 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035493.3 NP_001030570.1 8 - 9 1107 71
GenBank mRNA BC110166.1 - 8 - 9 1108 71
BT021887.1 - 8 - 9 1120 71

View Full Product Details