Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TYMS
Entrez Gene ID
507631
Gene Name
thymidylate synthetase
Gene Aliases
TS
Chromosome Location
Chr.24: 35553508 - 35564151 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037816.1 NP_001032905.1 6 - 7 898 103
GenBank mRNA BC111139.1 - 6 - 7 898 103

View Full Product Details