Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THUMPD3
Entrez Gene ID
508940
Gene Name
THUMP domain containing 3
Gene Aliases
-
Chromosome Location
Chr.22: 17218914 - 17239336 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038062.1 NP_001033151.1 2 - 3 332 127
GenBank mRNA BC111239.1 - 2 - 3 332 127
BT025397.1 - 2 - 3 376 127

View Full Product Details