Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGP1
Entrez Gene ID
539393
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
Gene Aliases
-
Chromosome Location
Chr.8: 59917058 - 59922872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038184.1 NP_001033273.1 11 - 12 1730 101
GenBank mRNA BC111331.1 - 11 - 12 1730 101

View Full Product Details