Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RGP1
Entrez Gene ID
539393
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
Gene Aliases
-
Chromosome Location
Chr.8: 59917058 - 59922872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038184.1 NP_001033273.1 4 - 5 645 100
GenBank mRNA BC111331.1 - 4 - 5 645 100

View Full Product Details