Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D2
Entrez Gene ID
539917
Gene Name
PIH1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.15: 22376627 - 22382689 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038189.2 NP_001033278.1 4 - 5 604 73
GenBank mRNA BC109576.1 - 4 - 5 605 73

View Full Product Details