Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN2
Entrez Gene ID
508601
Gene Name
centrin 2
Gene Aliases
-
Chromosome Location
Chr.X: 35129257 - 35133378 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038515.1 NP_001033604.1 3 - 4 313 152
GenBank mRNA BC109888.1 - 3 - 4 313 152

View Full Product Details