Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNASEH2A
Entrez Gene ID
512830
Gene Name
ribonuclease H2 subunit A
Gene Aliases
-
Chromosome Location
Chr.7: 12722240 - 12730210 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038534.2 NP_001033623.1 6 - 7 1078 78
GenBank mRNA BC109682.1 - 6 - 7 1079 78

View Full Product Details