The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038543.2 | NP_001033632.1 | 4 - 5 | 339 | 66 |
GenBank mRNA | BC109905.1 | - | 4 - 5 | 340 | 66 |